Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igf1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152968.1 | NP_694500.1 | 17 - 18 | 3290 | 57 | |
GenBank mRNA | AF400269.1 | - | 2 - 3 | 248 | 57 | |
AF400275.1 | - | 17 - 18 | 3290 | 57 | ||
BC163723.1 | - | 17 - 18 | 3354 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |