Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178304.3 | NP_840089.2 | 1 - 2 | 414 | 72 | |
GenBank mRNA | AY871211.1 | - | 1 - 2 | 414 | 72 | |
BC049439.1 | - | 1 - 2 | 208 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |