Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178304.3 | NP_840089.2 | 5 - 6 | 965 | 101 | |
GenBank mRNA | AB085761.1 | - | 5 - 6 | 657 | 101 | |
AY871211.1 | - | 5 - 6 | 965 | 101 | ||
BC049439.1 | - | 5 - 6 | 759 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |