Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183068.1 | NP_898891.1 | 4 - 5 | 729 | 77 | |
XM_005157501.4 | XP_005157558.1 | 5 - 6 | 1177 | 77 | ||
GenBank mRNA | AY183456.1 | - | 4 - 5 | 729 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Danio rerio Probe spans exons |