Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194385.1 | NP_919366.1 | 3 - 4 | 652 | 90 | |
GenBank mRNA | AY338730.1 | - | 3 - 4 | 652 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |