Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199659.2 | NP_955953.2 | 18 - 19 | 2284 | 98 | |
XM_021478841.1 | XP_021334516.1 | 18 - 19 | 2312 | 98 | ||
GenBank mRNA | BC066480.1 | - | 18 - 19 | 2284 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |