Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199659.2 | NP_955953.2 | 6 - 7 | 964 | 114 | |
XM_021478841.1 | XP_021334516.1 | 6 - 7 | 992 | 114 | ||
GenBank mRNA | BC045445.1 | - | 6 - 7 | 948 | 114 | |
BC066480.1 | - | 6 - 7 | 964 | 114 | ||
BC165664.1 | - | 5 - 6 | 663 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |