Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199659.2 | NP_955953.2 | 9 - 10 | 1416 | 144 | |
XM_021478841.1 | XP_021334516.1 | 9 - 10 | 1444 | 144 | ||
GenBank mRNA | BC045445.1 | - | 9 - 10 | 1400 | 144 | |
BC066480.1 | - | 9 - 10 | 1416 | 144 | ||
BC165664.1 | - | 8 - 9 | 1115 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |