Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnika | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199703.1 | NP_955997.1 | 7 - 8 | 925 | 62 | |
XM_005163387.4 | XP_005163444.1 | 7 - 8 | 956 | 62 | ||
XM_005163388.4 | XP_005163445.1 | 7 - 8 | 956 | 62 | ||
XM_005163389.4 | XP_005163446.1 | 7 - 8 | 956 | 62 | ||
XM_005163390.4 | XP_005163447.1 | 7 - 8 | 956 | 62 | ||
XM_005163391.4 | XP_005163448.1 | 7 - 8 | 956 | 62 | ||
XM_005163392.4 | XP_005163449.1 | 7 - 8 | 956 | 62 | ||
GenBank mRNA | BC055134.1 | - | 7 - 8 | 925 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |