Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199799.1 | NP_956093.1 | 6 - 7 | 998 | 79 | |
XM_005157506.4 | XP_005157563.1 | 6 - 7 | 952 | 79 | ||
XM_017359218.2 | XP_017214707.1 | 6 - 7 | 888 | 79 | ||
XM_021481342.1 | XP_021337017.1 | 7 - 8 | 966 | 79 | ||
GenBank mRNA | BC044362.1 | - | 6 - 7 | 998 | 79 | |
BC164349.1 | - | 5 - 6 | 781 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |