Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199805.1 | NP_956099.1 | 5 - 6 | 582 | 70 | |
XM_005158491.4 | XP_005158548.1 | 5 - 6 | 857 | 70 | ||
XM_005158492.4 | XP_005158549.1 | 5 - 6 | 729 | 70 | ||
XM_017351598.2 | XP_017207087.1 | 5 - 6 | 1101 | 70 | ||
GenBank mRNA | BC046874.1 | - | 5 - 6 | 582 | 70 | |
BC165673.1 | - | 4 - 5 | 499 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |