Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199805.1 | NP_956099.1 | 7 - 8 | 952 | 81 | |
XM_005158491.4 | XP_005158548.1 | 7 - 8 | 1227 | 81 | ||
XM_005158492.4 | XP_005158549.1 | 7 - 8 | 1099 | 81 | ||
XM_017351598.2 | XP_017207087.1 | 7 - 8 | 1471 | 81 | ||
GenBank mRNA | BC046874.1 | - | 7 - 8 | 952 | 81 | |
BC165673.1 | - | 6 - 7 | 869 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |