Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppiab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328424.1 | NP_001315353.1 | 3 - 4 | 383 | 124 | |
GenBank mRNA | AY391451.1 | - | 3 - 4 | 251 | 124 | |
BC059470.1 | - | 3 - 4 | 328 | 124 | ||
BC071370.1 | - | 3 - 4 | 260 | 124 | ||
CF998539.1 | - | 3 - 4 | 230 | 124 | ||
EH487851.1 | - | 3 - 4 | 320 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |