Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200304.1 | NP_956598.1 | 3 - 4 | 447 | 83 | |
GenBank mRNA | BC049525.1 | - | 3 - 4 | 447 | 83 | |
BC065428.1 | - | 3 - 4 | 317 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |