Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201343.2 | NP_958500.1 | 9 - 10 | 1269 | 69 | |
GenBank mRNA | AY398313.1 | - | 9 - 10 | 1267 | 69 | |
BC048036.1 | - | 9 - 10 | 1260 | 69 | ||
BC164199.1 | - | 8 - 9 | 1106 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |