Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205588.3 | NP_991151.3 | 3 - 4 | 486 | 64 | |
XM_021479142.1 | XP_021334817.1 | 4 - 5 | 566 | 64 | ||
GenBank mRNA | AF510720.1 | - | 3 - 4 | 64 | ||
BC094967.1 | - | 3 - 4 | 64 | |||
BC164351.1 | - | 3 - 4 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |