Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212713.1 | NP_997878.1 | 3 - 4 | 459 | 81 | |
XM_005167004.4 | XP_005167061.1 | 3 - 4 | 528 | 81 | ||
GenBank mRNA | BC046885.1 | - | 3 - 4 | 459 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |