Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212827.3 | NP_997992.2 | 6 - 7 | 690 | 67 | |
GenBank mRNA | AY185211.3 | - | 6 - 7 | 634 | 67 | |
BC070013.1 | - | 6 - 7 | 690 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |