Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212946.1 | NP_998111.2 | 4 - 5 | 1100 | 81 | |
GenBank mRNA | BC067591.1 | - | 4 - 5 | 1100 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |