Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scinlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213090.2 | NP_998255.2 | 3 - 4 | 407 | 99 | |
GenBank mRNA | BC066531.1 | - | 3 - 4 | 372 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |