Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213140.1 | NP_998305.1 | 4 - 5 | 468 | 64 | |
XM_005160655.3 | XP_005160712.1 | 4 - 5 | 512 | 64 | ||
GenBank mRNA | BC053254.1 | - | 4 - 5 | 468 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |