Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdha1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213393.1 | NP_998558.1 | 10 - 11 | 1135 | 70 | |
XM_005165134.4 | XP_005165191.1 | 11 - 12 | 1121 | 70 | ||
GenBank mRNA | BC060928.1 | - | 10 - 11 | 1048 | 70 | |
BC071373.1 | - | 10 - 11 | 1135 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |