Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdha1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213393.1 | NP_998558.1 | 4 - 5 | 549 | 66 | |
XM_005165134.4 | XP_005165191.1 | 5 - 6 | 535 | 66 | ||
GenBank mRNA | BC060928.1 | - | 4 - 5 | 462 | 66 | |
BC071373.1 | - | 4 - 5 | 549 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |