Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213529.2 | NP_998694.1 | 6 - 7 | 560 | 91 | |
XM_005157050.4 | XP_005157107.1 | 6 - 7 | 570 | 91 | ||
GenBank mRNA | BC044368.1 | - | 6 - 7 | 562 | 91 | |
BC165136.1 | - | 5 - 6 | 480 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |