Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328554.1 | NP_001315483.1 | 2 - 3 | 277 | 87 | |
NM_214713.2 | NP_999878.1 | 2 - 3 | 322 | 87 | ||
GenBank mRNA | BC052135.1 | - | 2 - 3 | 249 | 87 | |
BC066758.1 | - | 2 - 3 | 237 | 87 | ||
EH456078.1 | - | 2 - 3 | 270 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |