Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptenb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001822.2 | NP_001001822.1 | 3 - 4 | 812 | 119 | |
GenBank mRNA | AY398670.1 | - | 3 - 4 | 222 | 119 | |
AY398671.1 | - | 3 - 4 | 222 | 119 | ||
BC093110.1 | - | 3 - 4 | 819 | 119 | ||
BC107962.1 | - | 3 - 4 | 812 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |