Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002105.1 | NP_001002105.1 | 11 - 12 | 1182 | 65 | |
GenBank mRNA | BC071504.1 | - | 11 - 12 | 1182 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |