Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002226.1 | NP_001002226.1 | 4 - 5 | 647 | 115 | |
GenBank mRNA | BC074060.1 | - | 4 - 5 | 647 | 115 | |
BC164596.1 | - | 4 - 5 | 553 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |