Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002360.1 | NP_001002360.1 | 10 - 11 | 1146 | 86 | |
XM_005158270.4 | XP_005158327.1 | 10 - 11 | 964 | 86 | ||
GenBank mRNA | BC076052.1 | - | 10 - 11 | 1146 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |