Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002624.1 | NP_001002624.1 | 2 - 3 | 540 | 74 | |
GenBank mRNA | BC075960.1 | - | 2 - 3 | 540 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |