Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002666.1 | NP_001002666.2 | 2 - 3 | 353 | 70 | |
XM_021477460.1 | XP_021333135.1 | 2 - 3 | 492 | 70 | ||
GenBank mRNA | BC076553.1 | - | 2 - 3 | 353 | 70 | |
BC163973.1 | - | 2 - 3 | 267 | 70 | ||
HM138690.1 | - | 2 - 3 | 239 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |