Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002685.1 | NP_001002685.1 | 5 - 6 | 775 | 108 | |
GenBank mRNA | BC076515.1 | - | 5 - 6 | 775 | 108 | |
BC165872.1 | - | 4 - 5 | 618 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |