Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003489.1 | NP_001003489.1 | 3 - 4 | 455 | 60 | |
GenBank mRNA | BC078412.1 | - | 3 - 4 | 455 | 60 | |
BC150447.1 | - | 3 - 4 | 469 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |