Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003553.2 | NP_001003553.1 | 1 - 2 | 142 | 61 | |
GenBank mRNA | BC078284.1 | - | 1 - 2 | 147 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |