Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003619.2 | NP_001003619.1 | 8 - 9 | 983 | 61 | |
XM_009298030.3 | XP_009296305.1 | 5 - 6 | 597 | 61 | ||
XM_021470480.1 | XP_021326155.1 | 7 - 8 | 1501 | 61 | ||
GenBank mRNA | BC078313.1 | - | 8 - 9 | 986 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |