Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004654.1 | NP_001004654.1 | 1 - 2 | 169 | 82 | |
GenBank mRNA | BC081524.1 | - | 1 - 2 | 169 | 82 | |
BC164135.1 | - | 1 - 2 | 144 | 82 | ||
KT008425.1 | - | 1 - 2 | 110 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |