Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004654.1 | NP_001004654.1 | 3 - 4 | 457 | 84 | |
GenBank mRNA | BC081524.1 | - | 3 - 4 | 457 | 84 | |
BC164135.1 | - | 3 - 4 | 432 | 84 | ||
KT008425.1 | - | 3 - 4 | 398 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |