Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005604.1 | NP_001005604.1 | 1 - 2 | 67 | 69 | |
XM_005170982.4 | XP_005171039.1 | 4 - 5 | 1331 | 69 | ||
XM_005170983.4 | XP_005171040.1 | 5 - 6 | 1276 | 69 | ||
XM_005170984.4 | XP_005171041.1 | 4 - 5 | 1072 | 69 | ||
GenBank mRNA | AY296060.1 | - | 1 - 2 | 67 | 69 | |
BC171666.1 | - | 1 - 2 | 133 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |