Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005953.1 | NP_001005953.1 | 2 - 3 | 335 | 106 | |
GenBank mRNA | BC083448.1 | - | 2 - 3 | 335 | 106 | |
BC164224.1 | - | 1 - 2 | 187 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |