Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006103.2 | NP_001006103.2 | 3 - 4 | 582 | 73 | |
GenBank mRNA | BC083209.1 | - | 3 - 4 | 583 | 73 | |
BC164755.1 | - | 2 - 3 | 331 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |