Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014320.2 | NP_001014342.1 | 5 - 6 | 967 | 106 | |
XM_005173643.4 | XP_005173700.1 | 5 - 6 | 975 | 106 | ||
XM_005173644.4 | XP_005173701.1 | 5 - 6 | 994 | 106 | ||
GenBank mRNA | BC091786.1 | - | 5 - 6 | 969 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |