Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014320.2 | NP_001014342.1 | 10 - 11 | 2138 | 65 | |
XM_005173643.4 | XP_005173700.1 | 10 - 11 | 2161 | 65 | ||
XM_005173644.4 | XP_005173701.1 | 10 - 11 | 2180 | 65 | ||
GenBank mRNA | BC091786.1 | - | 10 - 11 | 2140 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |