Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017717.1 | NP_001017717.1 | 5 - 6 | 938 | 63 | |
XM_005168903.4 | XP_005168960.1 | 5 - 6 | 943 | 63 | ||
XM_005168904.4 | XP_005168961.1 | 5 - 6 | 937 | 63 | ||
XM_021477587.1 | XP_021333262.1 | 6 - 7 | 1039 | 63 | ||
GenBank mRNA | BC093246.1 | - | 5 - 6 | 938 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |