Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001023580.1 | NP_001018874.1 | 2 - 3 | 199 | 62 | |
GenBank mRNA | BC095899.1 | - | 2 - 3 | 199 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |