Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025554.2 | NP_001020725.1 | 8 - 9 | 1389 | 71 | |
GenBank mRNA | BC085624.1 | - | 7 - 8 | 1536 | 71 | |
BC124073.1 | - | 7 - 8 | 1530 | 71 | ||
BC134877.1 | - | 8 - 9 | 1389 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |