Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037237.1 | NP_001032314.1 | 11 - 12 | 1831 | 55 | |
XM_005163511.4 | XP_005163568.1 | 13 - 14 | 2016 | 55 | ||
XM_009298626.3 | XP_009296901.1 | 12 - 13 | 1991 | 55 | ||
XM_009298629.3 | XP_009296904.1 | 11 - 12 | 1934 | 55 | ||
XM_021480957.1 | XP_021336632.1 | 13 - 14 | 1956 | 55 | ||
XM_021480959.1 | XP_021336634.1 | 12 - 13 | 1890 | 55 | ||
GenBank mRNA | AY876015.1 | - | 11 - 12 | 1831 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |