Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039641.1 | NP_001034730.1 | 3 - 4 | 374 | 127 | |
GenBank mRNA | AY309262.1 | - | 3 - 4 | 374 | 127 | |
BC162305.1 | - | 3 - 4 | 346 | 127 | ||
BC162306.1 | - | 3 - 4 | 346 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |