Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030131.2 | NP_001025302.1 | 17 - 18 | 2434 | 86 | |
XM_005171778.4 | XP_005171835.1 | 18 - 19 | 2606 | 86 | ||
XM_005171779.4 | XP_005171836.1 | 18 - 19 | 2603 | 86 | ||
XM_021475966.1 | XP_021331641.1 | 16 - 17 | 2399 | 86 | ||
GenBank mRNA | BC115049.1 | - | 5 - 6 | 514 | 86 | |
BC133980.1 | - | 2 - 3 | 279 | 86 | ||
BC171681.1 | - | 17 - 18 | 2465 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |