Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040316.2 | NP_001035406.1 | 7 - 8 | 964 | 109 | |
GenBank mRNA | BC115252.1 | - | 7 - 8 | 965 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |