Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pitx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040346.2 | NP_001035436.3 | 2 - 3 | 467 | 61 | |
XM_021468902.1 | XP_021324577.1 | 3 - 4 | 428 | 61 | ||
GenBank mRNA | BC115352.1 | - | 2 - 3 | 419 | 61 | |
EU032338.1 | - | 2 - 3 | 467 | 61 | ||
EU580407.1 | - | 2 - 3 | 514 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |